Merck
  • Erythropoietin Intensifies the Proapoptotic Activity of LFM-A13 in Cells and in a Mouse Model of Colorectal Cancer.
Erythropoietin Intensifies the Proapoptotic Activity of LFM-A13 in Cells and in a Mouse Model of Colorectal Cancer.
International journal of molecular sciences (2018-04-25)
Anna Tankiewicz-Kwedlo, Justyna Magdalena Hermanowicz, Krystyna Pawlak, Robert Czarnomysy, Krzysztof Bielawski, Izabela Prokop, Dariusz Pawlak
ABSTRACT
The Bruton’s tyrosine kinase (BTK) inhibitor LFM-A13 has been widely employed as an antileukemic agent, but applications in solid cancer have been found recently. The compound promotes apoptosis, has an antiproliferative effect, and increases cancer cell sensitivity to chemotherapy drugs. We decided to assess the impact of the simultaneous use of erythropoietin (Epo) and LFM-A13 on signal transduction in colon DLD-1 and HT-29 cells, as well as in tumor xenografts. The induction of apoptosis by Epo and LFM-A-13 in the cells was confirmed by phosphatidylserine externalization, loss of mitochondrial membrane potential, and modulation of the expression of apoptotic protein BAX and antiapoptotic protein BCL-2 in colon adenocarcinoma cells. Nude mice were inoculated with adenocarcinoma cells and treated with Epo and LFM-A13 in order to evaluate the degree of tumor regression. The simultaneous use of Epo and LFM-A13 severely inhibited cell growth, activated apoptosis, and also inhibited tumor growth in xenografts. The addition of Epo to LFM-A13 intensified the antiproliferative effect of LFM-A13, confirmed by the loss of mitochondrial membrane potential and the accumulation of apoptotic colon cancer cells with externalized phosphatidylserine (PS). These preclinical results suggest that the combination of Epo and LFM-A13 has a high proapoptotic activity and should be tested in the clinic for the treatment of solid tumors such as colon cancer.
Sigma-Aldrich
Monoclonal Anti-BAX antibody produced in mouse, clone 1F5-1B7, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
JC-1, powder or solid (Crystals)
Sigma-Aldrich
Monoclonal Anti-Bcl-2 antibody produced in mouse, clone Bcl-2-100, ascites fluid
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
LFM-A13, ≥98% (HPLC), powder