Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Zirconium(IV) oxynitrate hydrate
Synonym(s):
Zirconyl nitrate hydrate
Linear Formula:
ZrO(NO
3
)
2
· xH
2
O
CAS Number:
14985-18-3
Molecular Weight:
231.23 (anhydrous basis)
EC Number:
237-529-3
Products
Brand
Sigma-Aldrich (2)
Color
Formula Weight
Physical Form
Purity
Chemical Composition
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
243493
Zirconium(IV) oxynitrate hydrate,
Expand
View Pricing
346462
Zirconium(IV) oxynitrate hydrate,
technical grade
Expand
View Pricing
380679
Zirconium(IV) oxynitrate hydrate,
99.99% trace metals basis
Expand
View Pricing