
Zidovudine Related Compound C

Synonym(s):
Thymine, 2,4-Dihydroxy-5-methylpyrimidine, 5-Methyluracil
Empirical Formula (Hill Notation):
C5H6N2O2
CAS Number:
Molecular Weight:
126.11
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing