Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Wright stain
Synonym(s):
Eosin Methylene blue according to Wright
CAS Number:
68988-92-1
Products
Product Category
stains (3)
Brand
Sigma-Aldrich (3)
Color
Physical Form
Product Line
Quality Segment
Application
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
45252
Wright stain,
BioReagent, suitable for microscopy
Expand
View Pricing
W0625
Wright stain,
≤4% purity (HPLC), powder
Expand
View Pricing
861375
Wright stain,
certified by the Biological Stain Commission
Expand
View Pricing