Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Wide mouth wash bottle
Products
Product Category
bottles (5)
Manufacturer
Capacity (mL)
Cap
Bottle material
Color
Vial Bottle Material
Vial Bottle Color
Package Size
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
HS120245
Wide mouth wash bottle,
white PE-LD bottle, capacity 500 mL
Expand
View Pricing
HS120249
Wide mouth wash bottle,
capacity 500 mL, blue PE-LD bottle
Expand
View Pricing
HS120248
Wide mouth wash bottle,
capacity 500 mL, green PE-LD bottle
Expand
View Pricing
HS120247
Wide mouth wash bottle,
capacity 500 mL, yellow PE-LD bottle
Expand
View Pricing
HS120246
Wide mouth wash bottle,
capacity 500 mL, red PE-LD bottle
Expand
View Pricing