Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Wheaton
®
Microliter Snap Caps With Septa
Products
Product Category
caps (22)
Cap
Color
Filter Products
Showing 1-20 of 22
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
DWK11-0053N
Wheaton
®
Microliter Snap Caps With Septa,
natural PTFE/silicone cap
Expand
View Pricing
DWK11-0053DB
Wheaton
®
Microliter Snap Caps With Septa,
dark blue PTFE/silicone cap
Expand
View Pricing
DWK11-0053K
Wheaton
®
Microliter Snap Caps With Septa,
black PTFE/silicone cap
Expand
View Pricing
DWK11-0054N
Wheaton
®
Microliter Snap Caps With Septa,
natural PTFE/silicone cap
Expand
View Pricing
DWK11-0054DB
Wheaton
®
Microliter Snap Caps With Septa,
dark blue PTFE/silicone cap
Expand
View Pricing
DWK11-0053R
Wheaton
®
Microliter Snap Caps With Septa,
red PTFE/silicone cap
Expand
View Pricing
DWK11-0054R
Wheaton
®
Microliter Snap Caps With Septa,
red PTFE/silicone cap
Expand
View Pricing
DWK11-0051B
Wheaton
®
Microliter Snap Caps With Septa,
light blue PTFE cap (Push fit)
Expand
View Pricing
DWK11-0056N
Wheaton
®
Microliter Snap Caps With Septa,
natural PTFE/silicone cap
Expand
View Pricing
DWK11-0051R
Wheaton
®
Microliter Snap Caps With Septa,
red PTFE cap (Push fit)
Expand
View Pricing
DWK11-0056B
Wheaton
®
Microliter Snap Caps With Septa,
light blue PTFE/silicone cap
Expand
View Pricing
DWK11-0055B
Wheaton
®
Microliter Snap Caps With Septa,
light blue PTFE/silicone/PTFE cap
Expand
View Pricing
DWK11-0056K
Wheaton
®
Microliter Snap Caps With Septa,
black PTFE/silicone cap
Expand
View Pricing
DWK11-0051G
Wheaton
®
Microliter Snap Caps With Septa,
green PTFE cap (Push fit)
Expand
View Pricing
DWK11-0051K
Wheaton
®
Microliter Snap Caps With Septa,
black PTFE cap (Push fit)
Expand
View Pricing
DWK11-0054G
Wheaton
®
Microliter Snap Caps With Septa,
green PTFE/silicone cap
Expand
View Pricing
DWK11-0055N
Wheaton
®
Microliter Snap Caps With Septa,
natural PTFE/silicone/PTFE cap
Expand
View Pricing
DWK11-0055R
Wheaton
®
Microliter Snap Caps With Septa,
red PTFE/silicone/PTFE cap
Expand
View Pricing
DWK11-0056DB
Wheaton
®
Microliter Snap Caps With Septa,
dark blue PTFE/silicone cap
Expand
View Pricing
DWK11-0054K
Wheaton
®
Microliter Snap Caps With Septa,
black PTFE/silicone cap
Expand
View Pricing
Page 1 of 2
Page 1 of 2