Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
WHEATON
®
amber sample vial with PTFE faced rubber lined cap packed in partitioned tray
Products
Product Category
vials (5)
Capacity (mL)
Diameter (mm)
Cap
Color
Package Size
Sterility
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
DWKW224604
WHEATON
®
amber sample vial with PTFE faced rubber lined cap packed in partitioned tray,
glass, tube capacity (20 mL), screw cap
Expand
View Pricing
DWKW224682
WHEATON
®
amber sample vial with PTFE faced rubber lined cap packed in partitioned tray,
glass, tube capacity (4 mL), screw cap
Expand
View Pricing
DWKW224605
WHEATON
®
amber sample vial with PTFE faced rubber lined cap packed in partitioned tray,
glass, tube capacity (40 mL), screw cap
Expand
View Pricing
DWKW224684
WHEATON
®
amber sample vial with PTFE faced rubber lined cap packed in partitioned tray,
glass, tube insert capacity (8 mL), screw cap
Expand
View Pricing
DWKW224681
WHEATON
®
amber sample vial with PTFE faced rubber lined cap packed in partitioned tray,
glass, tube capacity (2 mL), screw cap
Expand
View Pricing