Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Whatman
®
UniPrep
®
syringeless filters
Products
Product Category
filters (7)
Manufacturer
Capacity (mL)
Cap
Filter Capacity (ml)
Housing Material
Membrane Material
Pore size (µm)
Sterility
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
WHAUN113UORG
Whatman
®
UniPrep
®
syringeless filters,
PTFE membrane, pore size 0.45 μm
Expand
View Pricing
WHAUN513UAQU
Whatman
®
UniPrep
®
syringeless filters,
pore size 0.45 μm, PVDF membrane
Expand
View Pricing
WHAUN113UAQU
Whatman
®
UniPrep
®
syringeless filters,
PVDF membrane, pore size 0.45 μm
Expand
View Pricing
WHAUN113EORG
Whatman
®
UniPrep
®
syringeless filters,
PTFE membrane, pore size 0.2 μm
Expand
View Pricing
WHAUN113UGMF
Whatman
®
UniPrep
®
syringeless filters,
membrane (GMF), pore size 0.45 μm
Expand
View Pricing
WHAUN113UNYL
Whatman
®
UniPrep
®
syringeless filters,
Nylon membrane, pore size 0.45 μm
Expand
View Pricing
WHAUN113ENYL
Whatman
®
UniPrep
®
syringeless filters,
Nylon membrane, pore size 0.2 μm
Expand
View Pricing