Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Whatman
®
UNIFLO
®
25 syringe filters
Products
Product Category
syringe filters (14)
Manufacturer
Diameter (mm)
Surface Area cm²
Housing Material
Membrane Material
Package Size
Pore size (µm)
Sterility
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
WHA9913-2502
Whatman
®
UNIFLO
®
25 syringe filters,
PVDF membrane, sterile, pore size 0.22 μm, pack of 45 ea
Expand
View Pricing
WHA9914-2502
Whatman
®
UNIFLO
®
25 syringe filters,
polyethersulfone membrane, sterile, pore size 0.2 μm, pack of 45 ea
Expand
View Pricing
WHA9914-2504
Whatman
®
UNIFLO
®
25 syringe filters,
pore size 0.45 μm, polyethersulfone membrane, sterile, pack of 200 ea, pack of 45 ea
Expand
View Pricing
WHA9913-2504
Whatman
®
UNIFLO
®
25 syringe filters,
PVDF membrane, sterile, pore size 0.45 μm, pack of 45 ea
Expand
View Pricing
WHA99092502
Whatman
®
UNIFLO
®
25 syringe filters,
PVDF membrane, pore size 0.22 μm, pack of 500 ea
Expand
View Pricing
WHA99092504
Whatman
®
UNIFLO
®
25 syringe filters,
PVDF membrane, pore size 0.45 μm, pack of 500 ea
Expand
View Pricing
WHA9915-2502
Whatman
®
UNIFLO
®
25 syringe filters,
polyethersulfone membrane, sterile, pore size 0.2 μm, pack of 200 ea
Expand
View Pricing
WHA99102504
Whatman
®
UNIFLO
®
25 syringe filters,
Nylon membrane, pore size 0.45 μm, pack of 500 ea
Expand
View Pricing
WHA99102502
Whatman
®
UNIFLO
®
25 syringe filters,
Nylon membrane, pore size 0.22 μm, pack of 500 ea
Expand
View Pricing
WHA99112504
Whatman
®
UNIFLO
®
25 syringe filters,
PTFE membrane, pore size 0.45 μm, pack of 500 ea
Expand
View Pricing
WHA99112502
Whatman
®
UNIFLO
®
25 syringe filters,
PTFE membrane, pore size 0.22 μm, pack of 500 ea
Expand
View Pricing
WHA9915-2504
Whatman
®
UNIFLO
®
25 syringe filters,
polyethersulfone membrane, sterile, pore size 0.45 μm, pack of 200 ea
Expand
View Pricing
WHA99122502
Whatman
®
UNIFLO
®
25 syringe filters,
polyethersulfone membrane, pore size 0.45 μm, pack of 500 ea
Expand
View Pricing
WHA99122504
Whatman
®
UNIFLO
®
25 syringe filters,
polyethersulfone membrane, pore size 0.22 μm, pack of 500 ea
Expand
View Pricing