Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Whatman
®
qualitative filter paper for technical use, Grade Shark Skin
®
, creped
Products
Product Category
filter discs (14)
Manufacturer
Diameter (mm)
Package Size
Pore size (µm)
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
WHA10347522
Whatman
®
qualitative filter paper for technical use, Grade Shark Skin
®
, creped,
circles, diam. 340 mm, pack of 100
Expand
View Pricing
WHA10347511
Whatman
®
qualitative filter paper for technical use, Grade Shark Skin
®
, creped,
circles, diam. 125 mm, pack of 100
Expand
View Pricing
WHA10347509
Whatman
®
qualitative filter paper for technical use, Grade Shark Skin
®
, creped,
circles, diam. 90 mm, pack of 100
Expand
View Pricing
WHA10347521
Whatman
®
qualitative filter paper for technical use, Grade Shark Skin
®
, creped,
circles, diam. 270 mm, pack of 100
Expand
View Pricing
WHA10347512
Whatman
®
qualitative filter paper for technical use, Grade Shark Skin
®
, creped,
circles, diam. 185 mm, pack of 100
Expand
View Pricing
WHA10538877
Whatman
®
qualitative filter paper for technical use, Grade Shark Skin
®
, creped,
sheets, W × L 8 in. × 10 in., pack of 100
Expand
View Pricing
WHA10347530
Whatman
®
qualitative filter paper for technical use, Grade Shark Skin
®
, creped,
circles, diam. 320 mm, pack of 100
Expand
View Pricing
WHA10537138
Whatman
®
qualitative filter paper for technical use, Grade Shark Skin
®
, creped,
roll, W × L 21 in. × 750 ft, pack of 1
Expand
View Pricing
WHA10347519
Whatman
®
qualitative filter paper for technical use, Grade Shark Skin
®
, creped,
circles, diam. 240 mm, pack of 100
Expand
View Pricing
WHA10347510
Whatman
®
qualitative filter paper for technical use, Grade Shark Skin
®
, creped,
circles, diam. 110 mm, pack of 100
Expand
View Pricing
WHA10347525
Whatman
®
qualitative filter paper for technical use, Grade Shark Skin
®
, creped,
circles, diam. 500 mm, pack of 100
Expand
View Pricing
WHA10347513
Whatman
®
qualitative filter paper for technical use, Grade Shark Skin
®
, creped,
circles, diam. 150 mm, pack of 100
Expand
View Pricing
WHA10347577
Whatman
®
qualitative filter paper for technical use, Grade Shark Skin
®
, creped,
circles, diam. 290 mm, pack of 100
Expand
View Pricing
WHA10347523
Whatman
®
qualitative filter paper for technical use, Grade Shark Skin
®
, creped,
circles, diam. 385 mm, pack of 100
Expand
View Pricing