Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Whatman
®
filter paper, Grade 230, creped
Products
Product Category
filter discs (8)
Manufacturer
Diameter (mm)
Color
Package Size
Pore size (µm)
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
WHA5230185
Whatman
®
filter paper, Grade 230, creped,
circles, diam. 185 mm (7.3 in.), pkg of 50 ea
Expand
View Pricing
WHA5230110
Whatman
®
filter paper, Grade 230, creped,
circles, diam. 100 mm (3.9 in.), pkg of 50 ea
Expand
View Pricing
WHA5230090
Whatman
®
filter paper, Grade 230, creped,
circles, diam. 90 mm (3.5 in.), pkg of 50 ea
Expand
View Pricing
WHA5230250
Whatman
®
filter paper, Grade 230, creped,
circles, diam. 250 mm (9.8 in.), pkg of 50 ea
Expand
View Pricing
WHA5230150
Whatman
®
filter paper, Grade 230, creped,
circles, diam. 150 mm (5.9 in.), pkg of 50 ea
Expand
View Pricing
WHA5230125
Whatman
®
filter paper, Grade 230, creped,
circles, diam. 125 mm (4.9 in.), pkg of 50 ea
Expand
View Pricing
WHA5230330
Whatman
®
filter paper, Grade 230, creped,
circles, diam. 330 mm (13.0 in.), pkg of 50 ea
Expand
View Pricing
WHA5230240
Whatman
®
filter paper, Grade 230, creped,
circles, diam. 240 mm (9.4 in.), pkg of 50 ea
Expand
View Pricing
WHA5230500
Whatman
®
filter paper, Grade 230, creped,
circles, diam. 500 mm (19.7 in.), pkg of 50 ea
Expand
View Pricing