Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Whatman
®
filter paper, Grade 202, creped
Products
Product Category
filter discs (9)
Manufacturer
Diameter (mm)
Color
Package Size
Pore size (µm)
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
WHA5202125
Whatman
®
filter paper, Grade 202, creped,
circles, diam. 125 mm (4.9 in.), pkg of 100 ea
Expand
View Pricing
WHA5202200
Whatman
®
filter paper, Grade 202, creped,
circles, diam. 200 mm (7.9 in.), pkg of 100 ea
Expand
View Pricing
WHA5202250
Whatman
®
filter paper, Grade 202, creped,
circles, diam. 250 mm (9.8 in.), pkg of 100 ea
Expand
View Pricing
WHA5202090
Whatman
®
filter paper, Grade 202, creped,
circles, diam. 90 mm (3.5 in.), pkg of 100 ea
Expand
View Pricing
WHA5202185
Whatman
®
filter paper, Grade 202, creped,
circles, diam. 185 mm (7.3 in.), pkg of 100 ea
Expand
View Pricing
WHA5202110
Whatman
®
filter paper, Grade 202, creped,
circles, diam. 110 mm (4.3 in.), pkg of 100 ea
Expand
View Pricing
WHA5202150
Whatman
®
filter paper, Grade 202, creped,
circles, diam. 150 mm (5.9 in.), pkg of 100 ea
Expand
View Pricing
WHA5202330
Whatman
®
filter paper, Grade 202, creped,
circles, diam. 330 mm (13.0 in.), pkg of 100 ea
Expand
View Pricing
WHA5202240
Whatman
®
filter paper, Grade 202, creped,
circles, diam. 240 mm (9.4 in.), pkg of 100 ea
Expand
View Pricing