Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Water-
17
O
Synonym(s):
17
O Labeled water
Linear Formula:
H
2
17
O
CAS Number:
13768-40-6
Molecular Weight:
19.02
Products
Product Category
stable isotopes (13)
Brand
Sigma-Aldrich (13)
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Isotope
Filter Products
Showing 1-13 of 13
Compare
Product Number
Description
Pricing
602949
Water-
17
O,
20-24.9 atom %
17
O
Expand
View Pricing
609862
Water-
17
O,
90 atom %
17
O
Expand
View Pricing
603015
Water-
17
O,
10-14.9 atom %
17
O
Expand
View Pricing
602965
Water-
17
O,
40-44.9 atom %
17
O, 99% (CP)
Expand
View Pricing
603058
Water-
17
O,
70-75.9 atom %
17
O, 99% (CP)
Expand
View Pricing
603031
Water-
17
O,
45-49.9 atom %
17
O
Expand
View Pricing
603023
Water-
17
O,
25-29.9 atom %
17
O
Expand
View Pricing
618535
Water-
17
O,
80-84.9 atom %
17
O
Expand
View Pricing
602973
Water-
17
O,
30-34.9 atom %
17
O
Expand
View Pricing
645907
Water-
17
O,
7-9.9 atom %
17
O
Expand
View Pricing
603007
Water-
17
O,
35-39.9 atom %
17
O
Expand
View Pricing
603066
Water-
17
O,
75-80.9 atom %
17
O
Expand
View Pricing
602981
Water-
17
O,
15-19.9 atom %
17
O
Expand
View Pricing