Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Volac
®
volumetric flask
Products
Product Category
flasks (6)
Manufacturer
Capacity (mL)
Closure Type
Glassware Class
Package Size
Sterility
Style
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
Z306525
Volac
®
volumetric flask,
volume 500 mL, accuracy: 0.25 mL, certificate: no
Expand
View Pricing
Z306533
Volac
®
volumetric flask,
volume 500 mL, accuracy: 0.25 mL, certificate
Expand
View Pricing
Z306363
Volac
®
volumetric flask,
volume 10 mL, accuracy: 0.025 mL, certificate
Expand
View Pricing
Z306479
Volac
®
volumetric flask,
volume 100 mL, accuracy: 0.10 mL, certificate
Expand
View Pricing
Z306401
Volac
®
volumetric flask,
volume 20 mL, accuracy: 0.04 mL, certificate
Expand
View Pricing
Z306452
Volac
®
volumetric flask,
volume 50 mL, accuracy: 0.06 mL, certificate
Expand
View Pricing