Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Vials, screw top V-Vials
®
with open-top cap
Products
Product Category
vials (4)
Brand
Sigma-Aldrich (3)
Supelco (1)
Diameter (mm)
Height (mm)
Volume (ml)
Closure Type
Color
Vial Bottle Material
Vial Bottle Color
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
Z115150
Vials, screw top V-Vials
®
with open-top cap,
volume 5 mL, 20-400 thread, diam. × H 20 mm × 65 mm
Expand
View Pricing
Z115134
Vials, screw top V-Vials
®
with open-top cap,
volume 1 mL, 13-425 thread, diam. × H 13 mm × 44 mm
Expand
View Pricing
Z115142
Vials, screw top V-Vials
®
with open-top cap,
volume 3 mL, 20-400 thread, diam. × H 20 mm × 50 mm
Expand
View Pricing
Z115126
Vials, screw top V-Vials
®
with open-top cap,
volume 0.3 mL, 13-425 thread, diam. × H 13 mm × 35 mm
Expand
View Pricing