Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Vials, screw top, silane-treated
Products
Product Category
vials (6)
Brand
Supelco (6)
ID (mm)
OD (mm)
Volume (ml)
Closure Type
Color
Vial Bottle Material
Vial Bottle Color
Package Size
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
27217
Vials, screw top, silane-treated,
volume 4 mL, amber glass vial, O.D. × H × I.D. 15 mm × 45 mm × 8 mm, thread for 13-425
Expand
View Pricing
27238
Vials, screw top, silane-treated,
volume 2 mL, amber glass vial, O.D. × H × I.D. 12 mm × 32 mm × 4.6 mm
Expand
View Pricing
27114
Vials, screw top, silane-treated,
volume 4 mL, clear glass vial, O.D. × H × I.D. 15 mm × 45 mm × 8 mm
Expand
View Pricing
27029-U
Vials, screw top, silane-treated,
volume 2 mL, clear glass vial, O.D. × H × I.D. 12 mm × 32 mm × 4.6 mm, thread for 8-425
Expand
View Pricing
27219
Vials, screw top, silane-treated,
volume 2 mL, amber glass vial, O.D. × H × I.D. 12 mm × 32 mm × 4.6 mm, standard opening
Expand
View Pricing
27030
Vials, screw top, silane-treated,
volume 2 mL, clear glass vial, O.D. × H × I.D. 12 mm × 32 mm × 4.6 mm
Expand
View Pricing