Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Valco
®
Unions
Products
Brand
Supelco (1)
Manufacturer
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
22999
Valco
®
Unions,
internal reducing, bore 0.75 mm, for connecting 1/8 to 1/16 in.
Expand
View Pricing
59026
Valco
®
Unions,
internal reducing, for connecting 1/16 to 1/32 in., bore size 0.25 mm
Expand
View Pricing
58249
Valco
®
Unions,
internal reducing, 1/4 to 1/16 in., bore size 0.75 mm
Expand
View Pricing