Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Unwire
™
full racks, polyketone
Products
Product Category
racks (29)
Brand
Sigma-Aldrich (10)
Color
Height (mm)
Length (mm)
Package Size
Filter Products
Showing 1-20 of 29
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
R8391
Unwire
™
full racks, polyketone,
Holds 24 x 30-mm tubes, blue
Expand
View Pricing
R9016
Unwire
™
full racks, polyketone,
Holds 24 x 30-mm tubes, green
Expand
View Pricing
R6766
Unwire
™
full racks, polyketone,
orange, Holds 72 x 16-mm tubes
Expand
View Pricing
R8266
Unwire
™
full racks, polyketone,
Holds 40 x 25-mm tubes, blue
Expand
View Pricing
C1042
Unwire
™
full racks, polyketone,
Holds 24 x 30-mm tubes, red
Expand
View Pricing
R7766
Unwire
™
full racks, polyketone,
Holds 24 x 30-mm tubes, yellow
Expand
View Pricing
R5766
Unwire
™
full racks, polyketone,
Holds 72 x 16-mm tubes, red
Expand
View Pricing
R6266
Unwire
™
full racks, polyketone,
Holds 40 x 20-mm tubes, white
Expand
View Pricing
R8016
Unwire
™
full racks, polyketone,
Holds 72 x 16-mm tubes, blue
Expand
View Pricing
R6141
Unwire
™
full racks, polyketone,
Holds 72 x 16-mm tubes, white
Expand
View Pricing
R7016
Unwire
™
full racks, polyketone,
Holds 40 x 25-mm tubes, orange
Expand
View Pricing
C0917
Unwire
™
full racks, polyketone,
Holds 72 x 13-mm tubes, red
Expand
View Pricing
R6516
Unwire
™
full racks, polyketone,
Holds 24 x 30-mm tubes, white
Expand
View Pricing
R7891
Unwire
™
full racks, polyketone,
Holds 72 x 13-mm tubes, blue
Expand
View Pricing
R7391
Unwire
™
full racks, polyketone,
Holds 72 x 16-mm tubes, yellow
Expand
View Pricing
R6891
Unwire
™
full racks, polyketone,
orange, Holds 40 x 20-mm tubes
Expand
View Pricing
R6016
Unwire
™
full racks, polyketone,
Holds 72 x 13-mm tubes, white
Expand
View Pricing
R7141
Unwire
™
full racks, polyketone,
Holds 24 x 30-mm tubes, orange
Expand
View Pricing
R8141
Unwire
™
full racks, polyketone,
Holds 40 x 20-mm tubes, blue
Expand
View Pricing
R6641
Unwire
™
full racks, polyketone,
Holds 72 x 13-mm tubes, orange
Expand
View Pricing
Page 1 of 2
Page 1 of 2