Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Unsaturated Phosphatidylcholine Liposomes
Products
Brand
Sigma-Aldrich (40)
Chemical Composition
Quality Segment
Filter Products
Showing 1-20 of 40
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
CUP-512
Unsaturated Phosphatidylcholine Liposomes,
100% 16:0-18:1 PC (POPC)
Expand
View Pricing
CUP-506
Unsaturated Phosphatidylcholine Liposomes,
100% 18:1 (Delta-9-Cis) PC (DOPC)
Expand
View Pricing
CUP-526
Unsaturated Phosphatidylcholine Liposomes,
18:1 (Delta-9-Cis) PC (DOPC): Cholesterol (70:30 molar ratio)
Expand
View Pricing
CUP-532
Unsaturated Phosphatidylcholine Liposomes,
16:0-18:1 PC (POPC): Cholesterol (70:30 molar ratio)
Expand
View Pricing
CUP-501
Unsaturated Phosphatidylcholine Liposomes,
100% 14:1 (Delta-9-Cis) PC
Expand
View Pricing
CUP-517
Unsaturated Phosphatidylcholine Liposomes,
100% 18:0-20:4 PC
Expand
View Pricing
CUP-508
Unsaturated Phosphatidylcholine Liposomes,
100% 18:2 (Cis) PC (DLPC)
Expand
View Pricing
CUP-540
Unsaturated Phosphatidylcholine Liposomes,
18:1-18:0 PC: Cholesterol (70:30 molar ratio)
Expand
View Pricing
CUP-528
Unsaturated Phosphatidylcholine Liposomes,
18:2 (Cis) PC (DLPC): Cholesterol (70:30 molar ratio)
Expand
View Pricing
CUP-533
Unsaturated Phosphatidylcholine Liposomes,
16:0-18:2 PC: Cholesterol (70:30 molar ratio)
Expand
View Pricing
CUP-509
Unsaturated Phosphatidylcholine Liposomes,
100% 20:1 (Cis) PC
Expand
View Pricing
CUP-510
Unsaturated Phosphatidylcholine Liposomes,
100% 22:1 (Cis) PC
Expand
View Pricing
CUP-523
Unsaturated Phosphatidylcholine Liposomes,
16:1 (Delta-9-Cis) PC: Cholesterol (70:30 molar ratio)
Expand
View Pricing
CUP-534
Unsaturated Phosphatidylcholine Liposomes,
16:0-20:4 PC: Cholesterol (70:30 molar ratio)
Expand
View Pricing
CUP-502
Unsaturated Phosphatidylcholine Liposomes,
100% 14:1 (Delta-9-Trans) PC
Expand
View Pricing
CUP-504
Unsaturated Phosphatidylcholine Liposomes,
100% 16:1 (Delta-9-Trans) PC
Expand
View Pricing
CUP-507
Unsaturated Phosphatidylcholine Liposomes,
100% 18:1 (Delta-9-Trans) PC
Expand
View Pricing
CUP-514
Unsaturated Phosphatidylcholine Liposomes,
100% 16:0-20:4 PC
Expand
View Pricing
CUP-515
Unsaturated Phosphatidylcholine Liposomes,
100% 18:0-18:1 PC
Expand
View Pricing
CUP-516
Unsaturated Phosphatidylcholine Liposomes,
100% 18:0-18:2 PC
Expand
View Pricing
Page 1 of 2
Page 1 of 2