Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tungsten-rhenium alloy, W75Re25
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-20 of 46
Page 1 of 3
Page 1 of 3
Compare
Product Number
Description
Pricing
GF71114790
Tungsten-rhenium alloy, W75Re25,
2m wire, 0.05mm diameter, annealed
Expand
View Pricing
GF05533680
Tungsten-rhenium alloy, W75Re25,
10m wire, 0.1mm diameter, annealed
Expand
View Pricing
GF66886914
Tungsten-rhenium alloy, W75Re25,
1m wire, 0.2mm diameter, annealed
Expand
View Pricing
GF87054408
Tungsten-rhenium alloy, W75Re25,
0.1m wire, 0.178mm diameter, annealed
Expand
View Pricing
GF58623420
Tungsten-rhenium alloy, W75Re25,
0.1m wire, 0.075mm diameter, annealed
Expand
View Pricing
GF19034984
Tungsten-rhenium alloy, W75Re25,
0.5m wire, 0.75mm diameter, annealed
Expand
View Pricing
GF37843241
Tungsten-rhenium alloy, W75Re25,
0.1m wire, 0.1mm diameter, annealed
Expand
View Pricing
GF96113066
Tungsten-rhenium alloy, W75Re25,
0.5m wire, 0.05mm diameter, annealed
Expand
View Pricing
GF93822947
Tungsten-rhenium alloy, W75Re25,
0.5m wire, 1.0mm diameter, annealed
Expand
View Pricing
GF53009456
Tungsten-rhenium alloy, W75Re25,
0.05m wire, 1.0mm diameter, annealed
Expand
View Pricing
GF55955649
Tungsten-rhenium alloy, W75Re25,
0.2m wire, 0.75mm diameter, annealed
Expand
View Pricing
GF02357928
Tungsten-rhenium alloy, W75Re25,
0.1m wire, 0.75mm diameter, annealed
Expand
View Pricing
GF40905794
Tungsten-rhenium alloy, W75Re25,
1m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF64209786
Tungsten-rhenium alloy, W75Re25,
0.1m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF39127305
Tungsten-rhenium alloy, W75Re25,
5m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF60864712
Tungsten-rhenium alloy, W75Re25,
5m wire, 0.2mm diameter, annealed
Expand
View Pricing
GF50257087
Tungsten-rhenium alloy, W75Re25,
0.1m wire, 0.2mm diameter, annealed
Expand
View Pricing
GF56402052
Tungsten-rhenium alloy, W75Re25,
10m wire, 0.178mm diameter, annealed
Expand
View Pricing
GF27928266
Tungsten-rhenium alloy, W75Re25,
0.5m wire, 0.075mm diameter, annealed
Expand
View Pricing
GF06140629
Tungsten-rhenium alloy, W75Re25,
1m wire, 0.35mm diameter, annealed
Expand
View Pricing
Page 1 of 3
Page 1 of 3