Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tungsten-copper alloy, W72Cu28
Products
Manufacturer
Physical Form
Diameter (mm)
Length (mm)
Filter Products
Showing 1-19 of 19
Compare
Product Number
Description
Pricing
GF58496507
Tungsten-copper alloy, W72Cu28,
sheet, 50 x 50mm, 3.15mm thickness
Expand
View Pricing
GF99933924
Tungsten-copper alloy, W72Cu28,
115mm rod, 12.7mm diameter
Expand
View Pricing
GF58867970
Tungsten-copper alloy, W72Cu28,
sheet, 25 x 25mm, 3.15mm thickness
Expand
View Pricing
GF41561715
Tungsten-copper alloy, W72Cu28,
bar, 100mm x 25mm x 25mm
Expand
View Pricing
GF16195215
Tungsten-copper alloy, W72Cu28,
100mm rod, 3.5mm diameter
Expand
View Pricing
GF01840632
Tungsten-copper alloy, W72Cu28,
bar, 200mm x 25mm x 25mm
Expand
View Pricing
GF60332497
Tungsten-copper alloy, W72Cu28,
bar, 50mm x 25mm x 25mm
Expand
View Pricing
GF73172044
Tungsten-copper alloy, W72Cu28,
200mm rod, 6.25mm diameter
Expand
View Pricing
GF44202619
Tungsten-copper alloy, W72Cu28,
100mm rod, 8.0mm diameter
Expand
View Pricing
GF50185379
Tungsten-copper alloy, W72Cu28,
100mm rod, 6.25mm diameter
Expand
View Pricing
GF19572611
Tungsten-copper alloy, W72Cu28,
200mm rod, 16.0mm diameter
Expand
View Pricing
GF12063002
Tungsten-copper alloy, W72Cu28,
sheet, 50 x 100mm, 3.15mm thickness
Expand
View Pricing
GF46576660
Tungsten-copper alloy, W72Cu28,
100mm rod, 25mm diameter
Expand
View Pricing
GF22082655
Tungsten-copper alloy, W72Cu28,
200mm rod, 25mm diameter
Expand
View Pricing
GF85650846
Tungsten-copper alloy, W72Cu28,
200mm rod, 8.0mm diameter
Expand
View Pricing
GF24401483
Tungsten-copper alloy, W72Cu28,
200mm rod, 3.5mm diameter
Expand
View Pricing
GF07875142
Tungsten-copper alloy, W72Cu28,
sheet, 50 x 200mm, 3.15mm thickness
Expand
View Pricing
GF13249256
Tungsten-copper alloy, W72Cu28,
200mm rod, 12.7mm diameter
Expand
View Pricing
GF56156586
Tungsten-copper alloy, W72Cu28,
100mm rod, 16.0mm diameter
Expand
View Pricing