Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tungsten carbide/cobalt
Products
Manufacturer
Physical Form
Length (mm)
OD (mm)
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
GF55149396
Tungsten carbide/cobalt,
tube, WC 94%/Co 6%, outside diameter 1.0 mm, length 150 mm, wall thickness 0.3 mm
Expand
View Pricing
GF50243059
Tungsten carbide/cobalt,
sheet, WC 94%/Co 6%, thickness 2.0 mm, size 25 × 25 mm
Expand
View Pricing
GF07007116
Tungsten carbide/cobalt,
sheet, WC 94%/Co 6%, thickness 1.0 mm, size 100 × 100 mm
Expand
View Pricing
GF30067062
Tungsten carbide/cobalt,
rod, WC 94%/Co 6%, 2.0 mm diameter, length 150 mm
Expand
View Pricing
GF18976292
Tungsten carbide/cobalt,
rod, WC 94%/Co 6%, 0.5 mm diameter, length 30 mm
Expand
View Pricing
GF35160005
Tungsten carbide/cobalt,
sheet, WC 94%/Co 6%, thickness 5.0 mm, size 25 × 25 mm
Expand
View Pricing
GF02227980
Tungsten carbide/cobalt,
sphere, WC 94%/Co 6%, 0.40 mm diameter, 10_pcs, tolerance +/-2.5um
Expand
View Pricing
GF02536376
Tungsten carbide/cobalt,
rod, WC 94%/Co 6%, 2.0 mm diameter, length 100 mm
Expand
View Pricing
GF43667804
Tungsten carbide/cobalt,
rod, WC 94%/Co 6%, 6.35 mm diameter, length 150 mm
Expand
View Pricing
GF93674201
Tungsten carbide/cobalt,
tube, WC 94%/Co 6%, outside diameter 2.0 mm, wall thickness 0.6 mm, length 150 mm
Expand
View Pricing
GF68374222
Tungsten carbide/cobalt,
rod, WC 94%/Co 6%, 3.0 mm diameter, length 150 mm
Expand
View Pricing
GF70041192
Tungsten carbide/cobalt,
tube, WC 94%/Co 6%, outside diameter 0.5 mm, length 150 mm, wall thickness 0.2 mm
Expand
View Pricing
GF05507186
Tungsten carbide/cobalt,
sphere, WC 94%/Co 6%, 0.2 mm diameter, 10_pcs, tolerance +/-2.5um
Expand
View Pricing
GF13854911
Tungsten carbide/cobalt,
sphere, WC 94%/Co 6%, 0.35 mm diameter, 10_pcs, tolerance +/-2.5um
Expand
View Pricing
GF40731274
Tungsten carbide/cobalt,
sphere, WC 94%/Co 6%, 3.0 mm diameter, 10_pcs, tolerance +/-2.5um
Expand
View Pricing
GF12380329
Tungsten carbide/cobalt,
sphere, WC 94%/Co 6%, 6.0 mm diameter, 10_pcs, tolerance +/-2.5um
Expand
View Pricing