Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
TSKgel
®
ODS-100V (3 µm) HPLC Columns
Products
Product Category
columns (19)
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Technique
Filter Products
Showing 1-20 of 20
Compare
Product Number
Description
Pricing
821829
TSKgel
®
ODS-100V (3 µm) HPLC Columns,
L x I.D. 15 cm × 4.6 mm, C18 (octadecyl) phase
Expand
View Pricing
821997
TSKgel
®
ODS-100V (3 µm) HPLC Columns,
L x I.D. 1 cm × 2 mm, C18 (octadecyl) phase
Expand
View Pricing
821831
TSKgel
®
ODS-100V (3 µm) HPLC Columns,
L x I.D. 5 cm × 4.6 mm, C18 (octadecyl) phase
Expand
View Pricing
821940
TSKgel
®
ODS-100V (3 µm) HPLC Columns,
L x I.D. 10 cm × 4.6 mm, C18 (octadecyl) phase
Expand
View Pricing
821814
TSKgel
®
ODS-100V (3 µm) HPLC Columns,
L x I.D. 1 cm × 2 mm, C18 (octadecyl) phase
Expand
View Pricing
822701
TSKgel
®
ODS-100V (3 µm) HPLC Columns,
L x I.D. 25 cm × 2 mm, C18 (octadecyl) phase
Expand
View Pricing
821830
TSKgel
®
ODS-100V (3 µm) HPLC Columns,
L x I.D. 7.5 cm × 4.6 mm, C18 (octadecyl) phase
Expand
View Pricing
821810
TSKgel
®
ODS-100V (3 µm) HPLC Columns,
L x I.D. 15 cm × 2 mm, C18 (octadecyl) phase
Expand
View Pricing
822703
TSKgel
®
ODS-100V (3 µm) HPLC Columns,
L x I.D. 3.5 cm × 3 mm, C18 (octadecyl) phase
Expand
View Pricing
821812
TSKgel
®
ODS-100V (3 µm) HPLC Columns,
L x I.D. 5 cm × 2 mm, C18 (octadecyl) phase
Expand
View Pricing
821938
TSKgel
®
ODS-100V (3 µm) HPLC Columns,
L x I.D. 10 cm × 2 mm, C18 (octadecyl) phase
Expand
View Pricing
821838
TSKgel
®
ODS-100V (3 µm) HPLC Columns,
L x I.D. 3.5 cm × 1 mm, C18 (octadecyl) phase
Expand
View Pricing
822700
TSKgel
®
ODS-100V (3 µm) HPLC Columns,
L x I.D. 2 cm × 2 mm, C18 (octadecyl) phase
Expand
View Pricing
822704
TSKgel
®
ODS-100V (3 µm) HPLC Columns,
L x I.D. 25 cm × 3 mm, C18 (octadecyl) phase
Expand
View Pricing
821813
TSKgel
®
ODS-100V (3 µm) HPLC Columns,
L x I.D. 3.5 cm × 2 mm, C18 (octadecyl) phase
Expand
View Pricing
821811
TSKgel
®
ODS-100V (3 µm) HPLC Columns,
L x I.D. 7.5 cm × 2 mm, C18 (octadecyl) phase
Expand
View Pricing
822702
TSKgel
®
ODS-100V (3 µm) HPLC Columns,
L x I.D. 2 cm × 3 mm, C18 (octadecyl) phase
Expand
View Pricing
822706
TSKgel
®
ODS-100V (3 µm) HPLC Columns,
L x I.D. 3.5 cm × 4.6 mm, C18 (octadecyl) phase
Expand
View Pricing
822705
TSKgel
®
ODS-100V (3 µm) HPLC Columns,
L x I.D. 2 cm × 4.6 mm, C18 (octadecyl) phase
Expand
View Pricing
821839
TSKgel
®
ODS-100V (3 µm) HPLC Columns,
L x I.D. 5 cm × 1 mm, C18 (octadecyl) phase
Expand
View Pricing