Tryptic Soy Broth- Ready-to-use Bags, Tubes & Ampoules
Products
Product Category
Brand
Color
Physical Form
Agency
Chemical Composition
Capacity (mL)
Compatibility
Quality Segment
Closure Type
Application
Microorganism Suitability
Package Size
Sterility
Sterilization Method
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing