Skip to Content
Merck
Tryptic Soy Agar - Ready-to-use Settle Plates IsoBag®, DPTE Beta Bag with 190 mm Alpha Port, ICR plus lockable plate, sterile; γ-irradiated, plate diam. 90 mm, double packed of, suitable for air monitoring

Tryptic Soy Agar - Ready-to-use Settle Plates

Tryptic Soy Agar - Ready-to-use Settle Plates IsoBag®, DPTE Beta Bag with 190 mm Alpha Port, ICR plus lockable plate, sterile; γ-irradiated, plate diam. 90 mm, double packed of, suitable for air monitoring

Products

Product Category
Brand
Color
Physical Form
Agency
Chemical Composition
Diameter (mm)
Volume (ml)
Compatibility
Quality Segment
Application
Plate Material
Microorganism Suitability
Package Size
Sterility
Sterilization Method
Technique
Showing 1-20 of 23
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2