Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Trypsin solution from porcine pancreas
CAS Number:
9002-07-7
Products
Brand
Sigma-Aldrich (3)
Biological Source
pH Value
Physical Form
Product Line
Tissue
Quality Segment
Sterilization Method
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
T4549
Trypsin solution from porcine pancreas,
sterile-filtered, BioReagent, suitable for cell culture, 25 g porcine trypsin per liter in 0.9% sodium chloride
Expand
View Pricing
T4424
Trypsin solution from porcine pancreas,
1 ×, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin per liter in Hanks′ Balanced Salt Solution with phenol red
Expand
View Pricing
T4674
Trypsin solution from porcine pancreas,
sterile-filtered, BioReagent, suitable for cell culture, 25 g porcine trypsin per liter in Hanks′ Balanced Salt Solution with phenol red
Expand
View Pricing