Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Trypsin from bovine pancreas
CAS Number:
9002-07-7
EC Number:
232-650-8
Products
Product Category
proteases (7)
enzymes (1)
Brand
Sigma-Aldrich (8)
Biological Source
Physical Form
Agency
Tissue
Chemical Composition
Quality Segment
Application
Technique
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
T1426
Trypsin from bovine pancreas,
TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Expand
View Pricing
T8003
Trypsin from bovine pancreas,
Type I, ~10,000 BAEE units/mg protein
Expand
View Pricing
T9201
Trypsin from bovine pancreas,
powder, ≥7,500 BAEE units/mg solid
Expand
View Pricing
T8658
Trypsin from bovine pancreas,
suitable for protein sequencing, lyophilized powder
Expand
View Pricing
T8802
Trypsin from bovine pancreas,
TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Expand
View Pricing
T1005
Trypsin from bovine pancreas,
Type XI, lyophilized powder, ≥6,000 BAEE units/mg protein
Expand
View Pricing
T9935
Trypsin from bovine pancreas,
essentially salt-free, lyophilized powder, ≥9,000 BAEE units/mg protein, BioReagent, suitable for cell culture
Expand
View Pricing
T7309
Trypsin from bovine pancreas,
≥2,500 USP units/mg solid, meets USP testing specifications
Expand
View Pricing