Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Trypan Blue
Synonym(s):
Direct blue 14
Linear Formula:
C
34
H
24
N
6
O
14
S
4
Na
4
CAS Number:
72-57-1
Molecular Weight:
960.81
EC Number:
200-786-7
Products
Product Category
stains (1)
Brand
Sigma-Aldrich (2)
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Chemical Composition
Quality Segment
Application
Package Size
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
T6146
Trypan Blue,
powder, BioReagent, suitable for cell culture
Expand
View Pricing
302643
Trypan Blue,
Dye content 60 %, ≥80% (HPLC)
Expand
View Pricing