Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
TrueNorth
®
Flatpack PP Thin Film Freezer Boxes
Products
Product Category
boxes (3)
Color
Manufacturer
Diameter (mm)
Package Size
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
HS120372
TrueNorth
®
Flatpack PP Thin Film Freezer Boxes,
Holds 81 x 1.5-2 mL tubes, green, pk of 10
Expand
View Pricing
HS120370
TrueNorth
®
Flatpack PP Thin Film Freezer Boxes,
Holds 81 x 1.5-2 mL tubes, white, pk of 10
Expand
View Pricing
HS120371
TrueNorth
®
Flatpack PP Thin Film Freezer Boxes,
Holds 81 x 1.5-2 mL tubes, blue, pk of 10
Expand
View Pricing