
Tropisetron monohydrochloride

Synonym(s):
1H-Indole-3-carboxylic acid (3-endo)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester monohydrochloride, 3-Tropanylindole-3-carboxylate monohydrochloride, ICS-205,930, Navoban
Empirical Formula (Hill Notation):
C17H20N2O2 · HCl
CAS Number:
Molecular Weight:
320.81
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing