Skip to Content
Merck
Tris[2-phenylpyridinato-C2,N]iridium(III) sublimed grade

Tris[2-phenylpyridinato-C2,N]iridium(III)

Tris[2-phenylpyridinato-C2,N]iridium(III) sublimed grade
Synonym(s):
Ir(ppy)3, Iridium, tris[2-(2-pyridinyl-κN)phenyl-κC]
Empirical Formula (Hill Notation):
C33H24IrN3
CAS Number:
Molecular Weight:
654.78

Products

Product Category
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Special Grade
Showing 1-2 of 2