
Tris(2,2′-bipyridyl)dichlororuthenium(II) hexahydrate

Synonym(s):
Ru(BPY)3, Ruthenium-tris(2,2′-bipyridyl) dichloride, Tris(2,2′-bipyridyl)ruthenium(II) chloride hexahydrate
Empirical Formula (Hill Notation):
C30H24Cl2N6Ru · 6H2O
CAS Number:
Molecular Weight:
748.62
Products
Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Showing 1-2 of 2
Compare
Product Number
Description
Pricing