Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Trifluoroacetic anhydride
Synonym(s):
Trifluoroacetic anhydride, TFAA
Linear Formula:
(CF
3
CO)
2
O
CAS Number:
407-25-0
Molecular Weight:
210.03
EC Number:
206-982-9
Products
Brand
Sigma-Aldrich (1)
Supelco (1)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
8.08261
Trifluoroacetic anhydride,
for synthesis
Expand
View Pricing
106232
Trifluoroacetic anhydride,
ReagentPlus
®
, ≥99%
Expand
View Pricing
91719
Trifluoroacetic anhydride,
derivatization grade (GC derivatization), LiChropur
™
, ≥99.0% (GC)
Expand
View Pricing