Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Triethylammonium bicarbonate buffer
Synonym(s):
Triethylammonium hydrogen carbonate buffer
CAS Number:
15715-58-9
Products
Product Category
buffers (3)
Brand
Sigma-Aldrich (2)
Supelco (1)
Color
Feature
pH Value
Physical Form
Research Area
Quality Segment
Application
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
T7408
Triethylammonium bicarbonate buffer,
1.0 M, pH 8.5±0.1
Expand
View Pricing
90360
Triethylammonium bicarbonate buffer,
volatile buffer, ~1.0 M in H
2
O
Expand
View Pricing
18597
Triethylammonium bicarbonate buffer,
1 M, suitable for HPLC, LiChropur
™
Expand
View Pricing