Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tough-Tags
™
microcentrifuge tube labels
Products
Product Category
labels (15)
Color
Manufacturer
Length (in)
Color
Package Size
Technique
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
Z359122
Tough-Tags
™
microcentrifuge tube labels,
size 1.28 in. × 0.5 in., white
Expand
View Pricing
Z359106
Tough-Tags
™
microcentrifuge tube labels,
L × W 0.94 in. × 0.50 in., white
Expand
View Pricing
Z743680
Tough-Tags
™
microcentrifuge tube labels,
yellow, roll of 1000 labels, L × W 0.94 in. × 0.50 in.
Expand
View Pricing
Z743670
Tough-Tags
™
microcentrifuge tube labels,
green, roll of 1000 labels, L × W 0.94 in. × 0.50 in.
Expand
View Pricing
Z743675
Tough-Tags
™
microcentrifuge tube labels,
orange, roll of 1000 labels, L × W 0.94 in. × 0.50 in.
Expand
View Pricing
Z743679
Tough-Tags
™
microcentrifuge tube labels,
white, roll of 1000 labels, L × W 1.50 in. × 0.50 in.
Expand
View Pricing
Z743681
Tough-Tags
™
microcentrifuge tube labels,
yellow, roll of 1000 labels, L × W 1.28 in. × 0.50 in.
Expand
View Pricing
Z743669
Tough-Tags
™
microcentrifuge tube labels,
blue, roll of 1000 labels, L × W 1.28 in. × 0.50 in.
Expand
View Pricing
Z359114
Tough-Tags
™
microcentrifuge tube labels,
size 1.28 in. × 0.5 in., clear
Expand
View Pricing
Z743677
Tough-Tags
™
microcentrifuge tube labels,
red, roll of 1000 labels, L × W 0.94 in. × 0.50 in.
Expand
View Pricing
Z359092
Tough-Tags
™
microcentrifuge tube labels,
size 0.94 in. × 0.50 in., clear
Expand
View Pricing
Z743678
Tough-Tags
™
microcentrifuge tube labels,
red, roll of 1000 labels, L × W 1.28 in. × 0.50 in.
Expand
View Pricing
Z743668
Tough-Tags
™
microcentrifuge tube labels,
blue, roll of 1000 labels, L × W 0.94 in. × 0.50 in.
Expand
View Pricing
Z743676
Tough-Tags
™
microcentrifuge tube labels,
orange, roll of 1000 labels, L × W 1.28 in. × 0.50 in.
Expand
View Pricing
Z743671
Tough-Tags
™
microcentrifuge tube labels,
green, roll of 1000 labels, L × W 1.28 in. × 0.50 in.
Expand
View Pricing