Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tough-Spots
®
Synonym(s):
Tough-Spots
®
Products
Brand
Sigma-Aldrich (2)
Color
Manufacturer
Quality Segment
Color
Package Size
Technique
Filter Products
Showing 1-20 of 33
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
Z740255
Tough-Spots
®
,
size 3/8 in., white
Expand
View Pricing
Z743588
Tough-Spots
®
,
5 across, diam. 1/2 in., assorted colors, roll of 5000
Expand
View Pricing
Z688371
Tough-Spots
®
,
size diam. 1/2 in, white, pkg of 1000 roll
Expand
View Pricing
Z740261
Tough-Spots
®
,
size 1/2 in., white
Expand
View Pricing
Z743601
Tough-Spots
®
,
5 across, diam. 3/8 in., assorted colors, roll of 5000
Expand
View Pricing
Z740260
Tough-Spots
®
,
size 3/8 in., yellow
Expand
View Pricing
Z740256
Tough-Spots
®
,
size 3/8 in., blue
Expand
View Pricing
Z740257
Tough-Spots
®
,
size 3/8 in., green
Expand
View Pricing
Z740263
Tough-Spots
®
,
size 1/2 in., green
Expand
View Pricing
Z740259
Tough-Spots
®
,
size 3/8 in., red
Expand
View Pricing
Z740262
Tough-Spots
®
,
size 1/2 in., blue
Expand
View Pricing
Z740258
Tough-Spots
®
,
size 3/8 in., orange
Expand
View Pricing
Z740266
Tough-Spots
®
,
size 1/2 in., yellow
Expand
View Pricing
Z740264
Tough-Spots
®
,
size 1/2 in., orange
Expand
View Pricing
Z743611
Tough-Spots
®
,
diam. 7/16 in., white, roll of 1000
Expand
View Pricing
Z743609
Tough-Spots
®
,
diam. 3/8 in., pink, roll of 1000
Expand
View Pricing
Z743606
Tough-Spots
®
,
diam. 3/8 in., lavender, roll of 1000
Expand
View Pricing
Z743592
Tough-Spots
®
,
diam. 1/2 in., lavender, roll of 1000
Expand
View Pricing
Z740265
Tough-Spots
®
,
size 1/2 in., red
Expand
View Pricing
Z743595
Tough-Spots
®
,
diam. 1/2 in., pink, roll of 1000
Expand
View Pricing
Page 1 of 2
Page 1 of 2