Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tool steel
Products
Manufacturer
Format
Quality Segment
Reference Material Type
Special Grade
certified reference material (4)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
NIST1157
Tool steel,
NIST
®
SRM
®
1157
Expand
View Pricing
NIST1772
Tool steel,
NIST
®
SRM
®
1772, (S-7)
Expand
View Pricing
NIST132B
Tool steel,
NIST
®
SRM
®
132b, (AISI M2)
Expand
View Pricing
NIST1857
Tool steel,
NIST
®
SRM
®
1857, for abrasive wear standard
Expand
View Pricing