Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Titanium(IV) oxysulfate solution
Linear Formula:
TiOSO
4
CAS Number:
13825-74-6
Molecular Weight:
159.93
Products
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Analyte Suitability
Quality Segment
Application
Reagent Type
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
495379
Titanium(IV) oxysulfate solution,
~15 wt. % in dilute sulfuric acid, 99.99% trace metals basis
Expand
View Pricing
34244
Titanium(IV) oxysulfate solution,
1.9-2.1%, for determination of hydrogen peroxyde (H 15), according to DIN 38 409, part 15, DEV-18
Expand
View Pricing