Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Titanium-base alloy
Products
Manufacturer
Format
Quality Segment
Reference Material Type
Special Grade
certified reference material (6)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
NIST2433
Titanium-base alloy,
NIST
®
SRM
®
2433, 8Al - 1Mo - 1V
Expand
View Pricing
NIST648
Titanium-base alloy,
NIST
®
SRM
®
648, 5Al - 2Sn - 2Zr - 4Cr - 4Mo
Expand
View Pricing
NIST649
Titanium-base alloy,
NIST
®
SRM
®
649, 15V - 3Al - 3Cr - 3Sn
Expand
View Pricing
NIST2432
Titanium-base alloy,
NIST
®
SRM
®
2432, 10V - 2Fe - 3Al
Expand
View Pricing
NIST1128
Titanium-base alloy,
NIST
®
SRM
®
1128, 15V - 3Al - 3Cr - 3Sn
Expand
View Pricing
NIST2431
Titanium-base alloy,
NIST
®
SRM
®
2431, 6Al - 2Sn - 4Zr - 6Mo
Expand
View Pricing