Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Titanium-aluminum-vanadium alloy, Ti90Al6V4
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Length (mm)
Filter Products
Showing 1-20 of 83
Page 1 of 5
Page 1 of 5
Compare
Product Number
Description
Pricing
GF05987919
Titanium-aluminum-vanadium alloy, Ti90Al6V4,
5m wire, 1.0mm diameter, annealed
Expand
View Pricing
GF22378945
Titanium-aluminum-vanadium alloy, Ti90Al6V4,
sphere, 2mm diameter, 100pcs
Expand
View Pricing
GF21880638
Titanium-aluminum-vanadium alloy, Ti90Al6V4,
foil, 50 x 50mm, 0.1mm thickness, annealed
Expand
View Pricing
GF02815567
Titanium-aluminum-vanadium alloy, Ti90Al6V4,
powder, 450micron max. particle size, spherical powder, 50g
Expand
View Pricing
GF00557119
Titanium-aluminum-vanadium alloy, Ti90Al6V4,
foil, 300 x 300mm, 0.52mm thickness, annealed
Expand
View Pricing
GF30145272
Titanium-aluminum-vanadium alloy, Ti90Al6V4,
foil, 100 x 100mm, 1.0mm thickness, annealed
Expand
View Pricing
GF48059523
Titanium-aluminum-vanadium alloy, Ti90Al6V4,
foil, 25 x 25mm, 1.0mm thickness, annealed
Expand
View Pricing
GF04552185
Titanium-aluminum-vanadium alloy, Ti90Al6V4,
200mm rod, 7.0mm diameter
Expand
View Pricing
GF99149484
Titanium-aluminum-vanadium alloy, Ti90Al6V4,
powder, 45micron max. particle size, spherical powder, 50g
Expand
View Pricing
GF35079497
Titanium-aluminum-vanadium alloy, Ti90Al6V4,
foil, 100 x 100mm, 0.1mm thickness, annealed
Expand
View Pricing
GF40862493
Titanium-aluminum-vanadium alloy, Ti90Al6V4,
50mm rod, 10.0mm diameter
Expand
View Pricing
GF61352254
Titanium-aluminum-vanadium alloy, Ti90Al6V4,
powder, 45micron max. particle size, spherical powder, 200g
Expand
View Pricing
GF37182528
Titanium-aluminum-vanadium alloy, Ti90Al6V4,
foil, 50 x 50mm, 0.6mm thickness, annealed
Expand
View Pricing
GF56764053
Titanium-aluminum-vanadium alloy, Ti90Al6V4,
sphere, 6mm diameter, 100pcs
Expand
View Pricing
GF74920686
Titanium-aluminum-vanadium alloy, Ti90Al6V4,
powder, 45micron max. particle size, spherical powder, 100g
Expand
View Pricing
GF13109666
Titanium-aluminum-vanadium alloy, Ti90Al6V4,
1m wire, 0.10mm diameter, annealed
Expand
View Pricing
GF23173466
Titanium-aluminum-vanadium alloy, Ti90Al6V4,
sphere, 3mm diameter, 5.08μm sphericity, precision sphere, grade 200, 100pcs
Expand
View Pricing
GF07441163
Titanium-aluminum-vanadium alloy, Ti90Al6V4,
foil, 50 x 50mm, 1.0mm thickness, annealed
Expand
View Pricing
GF96516838
Titanium-aluminum-vanadium alloy, Ti90Al6V4,
foil, 50 x 50mm, 0.7mm thickness, annealed
Expand
View Pricing
GF27383014
Titanium-aluminum-vanadium alloy, Ti90Al6V4,
foil, 4mm disks, 0.30mm thickness, annealed
Expand
View Pricing
Page 1 of 5
Page 1 of 5