Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Titanium alloy beta 3, Ti77.5Mo12Zr6Sn4.5
Products
Manufacturer
Physical Form
Length (m)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
GF55233367
Titanium alloy beta 3, Ti77.5Mo12Zr6Sn4.5,
foil, 25 x 25mm, 0.42mm thickness, annealed, precipitation hardenable
Expand
View Pricing
GF72229440
Titanium alloy beta 3, Ti77.5Mo12Zr6Sn4.5,
foil, 0.2m coil, 0.42mm thickness, annealed, precipitation hardenable
Expand
View Pricing
GF65377839
Titanium alloy beta 3, Ti77.5Mo12Zr6Sn4.5,
foil, 150 x 150mm, 0.42mm thickness, annealed, precipitation hardenable
Expand
View Pricing
GF04170585
Titanium alloy beta 3, Ti77.5Mo12Zr6Sn4.5,
foil, 50 x 50mm, 0.42mm thickness, annealed, precipitation hardenable
Expand
View Pricing
GF21323023
Titanium alloy beta 3, Ti77.5Mo12Zr6Sn4.5,
foil, 100 x 100mm, 0.42mm thickness, annealed, precipitation hardenable
Expand
View Pricing
GF28681038
Titanium alloy beta 3, Ti77.5Mo12Zr6Sn4.5,
foil, 0.5m coil, 0.42mm thickness, annealed, precipitation hardenable
Expand
View Pricing