Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tinned copper alloy, CuSn
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
GF18004823
Tinned copper alloy, CuSn,
100m wire, 0.25mm diameter, annealed, pure sn coated
Expand
View Pricing
GF71179735
Tinned copper alloy, CuSn,
sn60/pb40 coated, foil, 0.5m coil, 15mm coil width, 0.05mm thickness, annealed
Expand
View Pricing
GF18368859
Tinned copper alloy, CuSn,
sn60/pb40 coated, foil, 5m coil, 15mm coil width, 0.05mm thickness, annealed
Expand
View Pricing
GF14070293
Tinned copper alloy, CuSn,
sn60/pb40 coated, foil, 10m coil, 15mm coil width, 0.05mm thickness, annealed
Expand
View Pricing
GF22787635
Tinned copper alloy, CuSn,
sn60/pb40 coated, foil, 1m coil, 15mm coil width, 0.05mm thickness, annealed
Expand
View Pricing