Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tin-lead-antimony alloy, Sn60Pb39Sb1
Products
Manufacturer
Physical Form
Length (m)
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
GF32892228
Tin-lead-antimony alloy, Sn60Pb39Sb1,
foil, 50 x 50mm, 0.006mm thickness, not light tested (nlt)
Expand
View Pricing
GF03278280
Tin-lead-antimony alloy, Sn60Pb39Sb1,
foil, 25 x 25mm, 0.006mm thickness, light tight (lt)
Expand
View Pricing
GF03193477
Tin-lead-antimony alloy, Sn60Pb39Sb1,
foil, 90 x 100mm, 0.10mm thickness
Expand
View Pricing
GF32960471
Tin-lead-antimony alloy, Sn60Pb39Sb1,
foil, 20 x 25mm, 0.10mm thickness
Expand
View Pricing
GF34627327
Tin-lead-antimony alloy, Sn60Pb39Sb1,
foil, 40 x 50mm, 0.10mm thickness
Expand
View Pricing
GF05089844
Tin-lead-antimony alloy, Sn60Pb39Sb1,
foil, 150 x 150mm, 0.006mm thickness, not light tested (nlt)
Expand
View Pricing
GF55067182
Tin-lead-antimony alloy, Sn60Pb39Sb1,
foil, 90 x 200mm, 0.10mm thickness
Expand
View Pricing
GF35382191
Tin-lead-antimony alloy, Sn60Pb39Sb1,
foil, 0.5m coil, 0.10mm thickness
Expand
View Pricing
GF19399607
Tin-lead-antimony alloy, Sn60Pb39Sb1,
foil, 300 x 300mm, 0.006mm thickness, not light tested (nlt)
Expand
View Pricing
GF36955705
Tin-lead-antimony alloy, Sn60Pb39Sb1,
foil, 100 x 100mm, 0.006mm thickness, not light tested (nlt)
Expand
View Pricing
GF04148681
Tin-lead-antimony alloy, Sn60Pb39Sb1,
foil, 1m coil, 0.10mm thickness
Expand
View Pricing
GF60898124
Tin-lead-antimony alloy, Sn60Pb39Sb1,
foil, 5m coil, 0.10mm thickness
Expand
View Pricing