Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Thoriated tungsten
Products
Manufacturer
Physical Form
Length (m)
Filter Products
Showing 1-17 of 17
Compare
Product Number
Description
Pricing
GF41206328
Thoriated tungsten,
wire, W 99%/Th 1%, 0.15 mm diameter, length 5 m, temper as drawn
Expand
View Pricing
GF70077764
Thoriated tungsten,
wire, 0.38 mm diameter, W 99%/Th 1%, length 5 m, temper annealed
Expand
View Pricing
GF45219922
Thoriated tungsten,
wire, W 99%/Th 1%, 1 mm diameter, length 0.2 m, temper annealed
Expand
View Pricing
GF92625161
Thoriated tungsten,
wire, W 99.4%/Th 0.6%, length 10 m, 0.025 mm diameter, temper annealed
Expand
View Pricing
GF47344079
Thoriated tungsten,
wire, W 99.4%/Th 0.6%, 0.013 mm diameter, length 10 m, temper annealed
Expand
View Pricing
GF85239502
Thoriated tungsten,
wire, 0.5 mm diameter, W 99%/Th 1%, length 1 m, temper annealed
Expand
View Pricing
GF23312225
Thoriated tungsten,
wire, W 99%/Th 1%, 0.5 mm diameter, length 2 m, temper annealed
Expand
View Pricing
GF01716690
Thoriated tungsten,
wire, W 99%/Th 1%, 0.38 mm diameter, length 2 m, temper annealed
Expand
View Pricing
GF71476533
Thoriated tungsten,
wire, W 99.4%/Th 0.6%, 0.025 mm diameter, length 200 m, temper annealed
Expand
View Pricing
GF71577434
Thoriated tungsten,
wire, 0.75 mm diameter, W 99%/Th 1%, length 0.5 m, temper annealed
Expand
View Pricing
GF30632071
Thoriated tungsten,
wire, W 99.4%/Th 0.6%, 0.025 mm diameter, length 25 m, temper annealed
Expand
View Pricing
GF88332420
Thoriated tungsten,
wire, W 99.4%/Th 0.6%, 0.05 mm diameter, length 50 m, temper annealed
Expand
View Pricing
GF35014766
Thoriated tungsten,
wire, W 99.4%/Th 0.6%, 1.0 mm diameter, length 0.5 m, temper annealed
Expand
View Pricing
GF47284296
Thoriated tungsten,
wire, W 99.4%/Th 0.6%, 0.013 mm diameter, length 25 m, temper annealed
Expand
View Pricing
GF47380754
Thoriated tungsten,
wire, W 99.4%/Th 0.6%, 0.013 mm diameter, length 200 m, temper annealed
Expand
View Pricing
GF41553491
Thoriated tungsten,
wire, W 99.4%/Th 0.6%, 0.05 mm diameter, length 100 m, temper annealed
Expand
View Pricing
GF94186547
Thoriated tungsten,
wire, 0.05 mm diameter, W 99.4%/Th 0.6%, length 200 m, temper annealed
Expand
View Pricing