Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Thionyl chloride
Synonym(s):
Thionyl chloride
Linear Formula:
SOCl
2
CAS Number:
7719-09-7
Molecular Weight:
118.97
EC Number:
231-748-8
Products
Product Category
salts (2)
Brand
Sigma-Aldrich (3)
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
8.08154
Thionyl chloride,
for synthesis
Expand
View Pricing
230464
Thionyl chloride,
ReagentPlus
®
, ≥99%
Expand
View Pricing
320536
Thionyl chloride,
reagent grade, 97%
Expand
View Pricing