Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tetrahexylammonium bromide
Linear Formula:
[CH
3
(CH
2
)
5
]
4
N(Br)
CAS Number:
4328-13-6
Molecular Weight:
434.58
EC Number:
224-363-1
Products
Brand
Sigma-Aldrich (1)
Supelco (1)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Puriss (1)
Puriss P.A. (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
252816
Tetrahexylammonium bromide,
99%
Expand
View Pricing
87297
Tetrahexylammonium bromide,
suitable for ion pair chromatography, LiChropur
™
, ≥99.0% (AT)
Expand
View Pricing