![Terbium lump, 25 mm max. lump size, weight 50 g, purity 99%](/deepweb/assets/sigmaaldrich/product/structures/425/786/12983bd0-28b5-4995-bc98-3dff094622cc/640/12983bd0-28b5-4995-bc98-3dff094622cc.png)
Terbium
![Terbium lump, 25 mm max. lump size, weight 50 g, purity 99%](/deepweb/assets/sigmaaldrich/product/structures/425/786/12983bd0-28b5-4995-bc98-3dff094622cc/640/12983bd0-28b5-4995-bc98-3dff094622cc.png)
Synonym(s):
Terbium
Empirical Formula (Hill Notation):
Tb
CAS Number:
Molecular Weight:
158.93
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Diameter (mm)
Length (mm)
Element
Particle Size (µm)
Showing 1-9 of 9
Compare
Product Number
Description
Pricing