Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tellurium
Products
Manufacturer
Physical Form
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
GF69312427
Tellurium,
powder, 75 max. part. size (micron), weight 100 g, purity 99.5%
Expand
View Pricing
GF48489430
Tellurium,
lump, 25 mm max. lump size, weight 100 g, high purity 99.999%
Expand
View Pricing
GF42954497
Tellurium,
powder, max. particle size 75 micron, weight 50 g, purity 99.5%
Expand
View Pricing
GF83536498
Tellurium,
powder, 75 max. part. size (micron), purity 99.5%, weight 500 g
Expand
View Pricing
GF61179217
Tellurium,
lump, 25 mm max. lump size, weight 200 g, high purity 99.999%
Expand
View Pricing
GF64052647
Tellurium,
powder, 75 max. part. size (micron), weight 200 g, purity 99.5%
Expand
View Pricing
GF59552185
Tellurium,
lump, 25 mm max. lump size, weight 50 g, purity 99.95%
Expand
View Pricing
GF23587915
Tellurium,
lump, 25 mm max. lump size, weight 50 g, high purity 99.999%
Expand
View Pricing
GF44023193
Tellurium,
lump, 25 mm max. lump size, weight 100 g, purity 99.95%
Expand
View Pricing
GF02899828
Tellurium,
lump, 25 mm max. lump size, weight 200 g, purity 99.95%
Expand
View Pricing