Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tantalum-tungsten alloy, Ta90W10
Products
Manufacturer
Physical Form
Diameter (mm)
Length (mm)
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
GF34123766
Tantalum-tungsten alloy, Ta90W10,
100mm rod, 1.5mm diameter, annealed
Expand
View Pricing
GF67833568
Tantalum-tungsten alloy, Ta90W10,
500mm rod, 8.0mm diameter, annealed
Expand
View Pricing
GF24550118
Tantalum-tungsten alloy, Ta90W10,
100mm rod, 8.0mm diameter, annealed
Expand
View Pricing
GF94353363
Tantalum-tungsten alloy, Ta90W10,
1000mm rod, 1.5mm diameter, annealed
Expand
View Pricing
GF35935511
Tantalum-tungsten alloy, Ta90W10,
500mm rod, 1.5mm diameter, annealed
Expand
View Pricing
GF11520043
Tantalum-tungsten alloy, Ta90W10,
foil, 100 x 100mm, 0.25mm thickness
Expand
View Pricing
GF61515895
Tantalum-tungsten alloy, Ta90W10,
200mm rod, 10.0mm diameter, annealed
Expand
View Pricing
GF21603709
Tantalum-tungsten alloy, Ta90W10,
foil, 50 x 50mm, 0.25mm thickness
Expand
View Pricing
GF54945502
Tantalum-tungsten alloy, Ta90W10,
100mm rod, 5.0mm diameter, annealed
Expand
View Pricing
GF46526205
Tantalum-tungsten alloy, Ta90W10,
50mm rod, 5.0mm diameter, annealed
Expand
View Pricing
GF22008507
Tantalum-tungsten alloy, Ta90W10,
200mm rod, 1.5mm diameter, annealed
Expand
View Pricing
GF07448889
Tantalum-tungsten alloy, Ta90W10,
foil, 50 x 50mm, 0.50mm thickness
Expand
View Pricing
GF25439643
Tantalum-tungsten alloy, Ta90W10,
200mm rod, 8.0mm diameter, annealed
Expand
View Pricing
GF90674051
Tantalum-tungsten alloy, Ta90W10,
100mm rod, 10.0mm diameter, annealed
Expand
View Pricing