Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
T1 - thermocouple alloy, Ni90Cr10
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Length (mm)
Filter Products
Showing 1-20 of 113
Page 1 of 6
Page 1 of 6
Compare
Product Number
Description
Pricing
GF20450405
T1 - thermocouple alloy, Ni90Cr10,
rod, 100mm x 12mm diameter, as drawn
Expand
View Pricing
GF14061428
T1 - thermocouple alloy, Ni90Cr10,
wire, Ni 90%/Cr 10%, 0.01 mm diameter, length 10 m, temper annealed
Expand
View Pricing
GF93293837
T1 - thermocouple alloy, Ni90Cr10,
100m wire, 0.025mm diameter, annealed
Expand
View Pricing
GF16029480
T1 - thermocouple alloy, Ni90Cr10,
rod, 100mm x 9mm diameter, as drawn
Expand
View Pricing
GF18835184
T1 - thermocouple alloy, Ni90Cr10,
rod, 1000mm x 2.0mm diameter, annealed
Expand
View Pricing
GF92077027
T1 - thermocouple alloy, Ni90Cr10,
rod, 500mm x 2.0mm diameter, annealed
Expand
View Pricing
GF05254635
T1 - thermocouple alloy, Ni90Cr10,
rod, 100mm x 2.0mm diameter, annealed
Expand
View Pricing
GF20817198
T1 - thermocouple alloy, Ni90Cr10,
insulated wire, 10m length, 0.05mm conductor diameter, 0.005mm insulation thickness, polyurethane insulation
Expand
View Pricing
GF12420706
T1 - thermocouple alloy, Ni90Cr10,
10m wire, 0.05mm diameter, annealed
Expand
View Pricing
GF22980483
T1 - thermocouple alloy, Ni90Cr10,
insulated wire, 100m length, 0.25mm conductor diameter, 0.018mm insulation thickness, polyester insulation
Expand
View Pricing
GF98469747
T1 - thermocouple alloy, Ni90Cr10,
rod, 200mm x 2.0mm diameter, annealed
Expand
View Pricing
GF82566535
T1 - thermocouple alloy, Ni90Cr10,
insulated wire, 5m length, 0.05mm conductor diameter, 0.005mm insulation thickness, polyurethane insulation
Expand
View Pricing
GF35745835
T1 - thermocouple alloy, Ni90Cr10,
1m wire, 1.0mm diameter, annealed
Expand
View Pricing
GF66191610
T1 - thermocouple alloy, Ni90Cr10,
500m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF39237857
T1 - thermocouple alloy, Ni90Cr10,
foil, 0.2m coil, 38mm coil width, 0.60mm thickness, annealed
Expand
View Pricing
GF98219885
T1 - thermocouple alloy, Ni90Cr10,
foil, 0.2m coil, 100mm coil width, 0.50mm thickness, annealed
Expand
View Pricing
GF99982265
T1 - thermocouple alloy, Ni90Cr10,
foil, 0.1m coil, 100mm coil width, 0.25mm thickness, annealed
Expand
View Pricing
GF45167564
T1 - thermocouple alloy, Ni90Cr10,
rod, 200mm x 12mm diameter, as drawn
Expand
View Pricing
GF44336875
T1 - thermocouple alloy, Ni90Cr10,
rod, 500mm x 6mm diameter, as drawn
Expand
View Pricing
GF93954695
T1 - thermocouple alloy, Ni90Cr10,
rod, 200mm x 6mm diameter, as drawn
Expand
View Pricing
Page 1 of 6
Page 1 of 6